missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromophenetole, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 199081000

 View more versions of this product

Catalog No. AC199081000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Bromophenetole
588-96-5
100.0
Glass bottle
Clear colorless to yellow
100g
97%
BrC6H4OC2H5
MFCD00000098
10, 1400
97%
96.0
Authentic
1.5500 to 1.5520 (20°C, 589nm)
Liquid
≥96.0%
C8H9BrO
201.06
06, 199
4-Bromophenyl ethyl ether

Chemical Identifiers

588-96-5
100.0
C8H9BrO
201.06
4-Bromophenyl ethyl ether
≥96.0%
96.0
97%
BrC6H4OC2H5
MFCD00000098
4-Bromophenetole, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 209-629-7

TSCA : TSCA