Please login to your online account to display your discounted pricing

4-Bromophenetole, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Bromophenetole
Name Note 97%
CAS 588-96-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H9BrO
Linear Formula BrC6H4OC2H5
Formula Weight 201.06
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC199080050

  SDS
thermo fisher scientific
199080050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC199081000

  SDS
thermo fisher scientific
199081000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 201.06
Physical Form Liquid
Quantity 5g
Color Clear colorless to yellow
TSCA TSCA
EINECS Number 209-629-7
Beilstein 06, 199
Merck Index 10, 1400
CAS Min % 96.0
CAS Max % 100.0
Synonym 4-Bromophenyl ethyl ether
Molecular Formula C8H9BrO
Linear Formula BrC6H4OC2H5
MDL Number MFCD00000098
Chemical Name or Material 4-Bromophenetole
CAS 588-96-5
Assay Percent Range 97%
Name Note 97%
Infrared Spectrum Authentic
Refractive Index 1.5500 to 1.5520 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.