missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromophenethyl alcohol, 99%, ACROS Organics™

Chemical Identifiers

CAS 4654-39-1
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00002897
InChI Key PMOSJSPFNDUAFY-UHFFFAOYSA-N
Synonym 2-4-bromo-phenyl-ethanol, 2-4-bromophenyl ethan-1-ol, 2-4-bromophenyl ethanol, 2-4-bromophenyl ethyl alcohol, 2-p-bromophenyl ethanol, 4-bromobenzene ethanol, 4-bromophenethyl alcohol, benzeneethanol, 4-bromo, p-bromophenethyl alcohol, phenethyl alcohol, p-bromo
PubChem CID 72851
IUPAC Name 2-(4-bromophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC179590100
SDS View Documents
Thermo Fisher Scientific
179590100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

4654-39-1
201.06
PMOSJSPFNDUAFY-UHFFFAOYSA-N
72851
C1=CC(=CC=C1CCO)Br
C8H9BrO
MFCD00002897
2-4-bromo-phenyl-ethanol, 2-4-bromophenyl ethan-1-ol, 2-4-bromophenyl ethanol, 2-4-bromophenyl ethyl alcohol, 2-p-bromophenyl ethanol, 4-bromobenzene ethanol, 4-bromophenethyl alcohol, benzeneethanol, 4-bromo, p-bromophenethyl alcohol, phenethyl alcohol, p-bromo
2-(4-bromophenyl)ethanol

Specifications

1.4300g/mL
Authentic
1.5720 to 1.574
96°C to 99°C (0.5mmHg)
10g
98.5
99%
BrC6H4CH2CH2OH
06, III, 1713
PMOSJSPFNDUAFY-UHFFFAOYSA-N
2-(4-bromophenyl)ethanol
72851
Liquid
4-Bromophenethyl alcohol, 99%
>110°C
Glass bottle
1.43
Colorless to Yellow
4654-39-1
100.0
C8H9BrO
MFCD00002897
2-4-bromo-phenyl-ethanol, 2-4-bromophenyl ethan-1-ol, 2-4-bromophenyl ethanol, 2-4-bromophenyl ethyl alcohol, 2-p-bromophenyl ethanol, 4-bromobenzene ethanol, 4-bromophenethyl alcohol, benzeneethanol, 4-bromo, p-bromophenethyl alcohol, phenethyl alcohol, p-bromo
C1=CC(=CC=C1CCO)Br
201.06
201.06
99%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-093-7

SDS