Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromonaphthalene-1-boronic acid, 97%

Chemical Identifiers

Chemical Name or Material 4-Bromonaphthalene-1-boronic acid
CAS 145965-14-6
Formula Weight 250.88
MDL Number MFCD08701764
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5562503

 
alfa aesar™
H5562503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5562506

 
alfa aesar™
H5562506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55625MD

 
alfa aesar™
H55625MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 230° to 260°C
Formula Weight 250.88
Chemical Name or Material 4-Bromonaphthalene-1-boronic acid
Color White to pale brown
Physical Form Solid
Quantity 1g
Assay 97%
MDL Number MFCD08701764
CAS 145965-14-6