missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromonaphthalene-1-boronic acid, 97%

Manufacturer:  Alfa Aesar™ H55625MD

 View more versions of this product

Catalog No. AAH55625MD



Specifications

Specifications

4-Bromonaphthalene-1-boronic acid
230°C to 260°C
145965-14-6
MFCD08701764
DKKOMUVRCPKTFX-UHFFFAOYSA-N
(4-bromonaphthalen-1-yl)boronic acid
23005424
Solid
Brown to White
250mg
C10H8BBrO2
1-bromo-4-dihydroxyboryl naphthalene
B(C1=CC=C(C2=CC=CC=C12)Br)(O)O
250.886
250.88
97%

Chemical Identifiers

145965-14-6
250.886
DKKOMUVRCPKTFX-UHFFFAOYSA-N
23005424
B(C1=CC=C(C2=CC=CC=C12)Br)(O)O
C10H8BBrO2
MFCD08701764
1-bromo-4-dihydroxyboryl naphthalene
(4-bromonaphthalen-1-yl)boronic acid
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents