Please login to your online account to display your discounted pricing

4-(Bromomethyl)tetrahydropyran, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific uk ltd  CC29908DA

 View more versions of this product

Catalog No. CC29908DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 179.06
Quantity 1g
Assay 97%
Chemical Name or Material 4-(Bromomethyl)tetrahydropyran
CAS 125552-89-8
Molecular Formula C6H11BrO
Packaging Amber Glass Bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material 4-(Bromomethyl)tetrahydropyran
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H11BrO
Formula Weight 179.06