Please login to your online account to display your discounted pricing

4-(Bromomethyl)pyridine hydrobromide, 97%


Manufacturer: thermo fisher scientific  AC397330050

 View more versions of this product

Catalog No. AC397330050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 252.93
Quantity 5g
Chemical Name or Material 4-(Bromomethyl)pyridine hydrobromide
Molecular Formula C6H6BrN·HBr
Packaging Glass bottle
CAS 73870-24-3
Assay Percent Range 99%

Specifications

Chemical Name or Material 4-(Bromomethyl)pyridine hydrobromide
CAS 73870-24-3
Molecular Formula C6H6BrN·HBr
Formula Weight 252.93