missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Bromomethyl)phenylacetic Acid 98.0+%, TCI America™
Supplier: TCI America B135125G
Specifications
4-(Bromomethyl)phenylacetic Acid | |
180°C | |
C9H9BrO2 | |
25 g | |
WCOCCXZFEJGHTC-UHFFFAOYSA-N | |
2-[4-(bromomethyl)phenyl]acetic acid | |
4519056 | |
≥98.0% (T) |
13737-36-5 | |
White | |
MFCD00010632 | |
4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid | |
C1=CC(=CC=C1CC(=O)O)CBr | |
229.073 | |
229.07 | |
Crystalline Powder |
Chemical Identifiers
13737-36-5 | |
229.073 | |
WCOCCXZFEJGHTC-UHFFFAOYSA-N | |
4519056 | |
C1=CC(=CC=C1CC(=O)O)CBr |
C9H9BrO2 | |
MFCD00010632 | |
4-bromomethyl phenylacetic acid, 4-bromomethylphenylacetic acid, 2-4-bromomethyl phenyl acetic acid, pam acid, br-pam-linker, 4-carboxymethyl benzyl bromide, 4-bromomethyl phenyl acetic acid, 4-bromomethyl phenylaceticacid | |
2-[4-(bromomethyl)phenyl]acetic acid |
Safety and Handling
TSCA : No