missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%

Manufacturer:  Alfa Aesar™ H5539414

 View more versions of this product

Catalog No. AAH5539414



Specifications

Specifications

4'-(Bromomethyl)biphenyl-2-carbonitrile
25g
C14H10BrN
UN3439
LFFIEVAMVPCZNA-UHFFFAOYSA-N
2-[4-(bromomethyl)phenyl]benzonitrile
1501912
98%
125°C to 128°C
114772-54-2
MFCD00671503
2'-cyano-4-bromomethylbiphenyl
C1=CC=C(C(=C1)C#N)C2=CC=C(C=C2)CBr
272.145
272.14

Chemical Identifiers

114772-54-2
272.145
LFFIEVAMVPCZNA-UHFFFAOYSA-N
1501912
C1=CC=C(C(=C1)C#N)C2=CC=C(C=C2)CBr
C14H10BrN
MFCD00671503
2'-cyano-4-bromomethylbiphenyl
2-[4-(bromomethyl)phenyl]benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents