missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%

Manufacturer:  Alfa Aesar™ H5539403

 View more versions of this product

Catalog No. AAH5539403



Specifications

Specifications

4'-(Bromomethyl)biphenyl-2-carbonitrile
1g
C14H10BrN
UN3439
LFFIEVAMVPCZNA-UHFFFAOYSA-N
2-[4-(bromomethyl)phenyl]benzonitrile
1501912
98%
125°C to 128°C
114772-54-2
MFCD00671503
2'-cyano-4-bromomethylbiphenyl
C1=CC=C(C(=C1)C#N)C2=CC=C(C=C2)CBr
272.145
272.14

Chemical Identifiers

114772-54-2
272.145
LFFIEVAMVPCZNA-UHFFFAOYSA-N
1501912
C1=CC=C(C(=C1)C#N)C2=CC=C(C=C2)CBr
C14H10BrN
MFCD00671503
2'-cyano-4-bromomethylbiphenyl
2-[4-(bromomethyl)phenyl]benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 425-280-5

TSCA : No

Recommended Storage : Ambient temperatures

Documents