Please login to your online account to display your discounted pricing

Alfa Aesar™ [4-(Bromomethyl)benzyl]triphenylphosphonium bromide, 98%

$82.50 - $1,186.78

Chemical Identifiers

Chemical Name or Material [4-(Bromomethyl)benzyl]triphenylphosphonium bromide
CAS 14366-74-6
Molecular Formula C26H23Br2P
Formula Weight 526.25
MDL Number MFCD00031635
Beilstein 3820074
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1756314

  SDS
alfa aesar™
A1756314
25g Each for $82.50

AAA1756322

  SDS
alfa aesar™
A1756322
100g Each for $278.61

AAA1756336

  SDS
alfa aesar™
A1756336
500g Each for $1,186.78
Description & Specifications

Specifications

Formula Weight 526.25
Chemical Name or Material [4-(Bromomethyl)benzyl]triphenylphosphonium bromide
Assay 98%
Molecular Formula C26H23Br2P
Melting Point 246° to 248°C (dec.)
Quantity 25g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number TA1854000
MDL Number MFCD00031635
CAS 14366-74-6
Beilstein 3820074
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.