Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Bromomethyl)benzeneboronic acid pinacol ester, 95%

Chemical Identifiers

Chemical Name or Material 4-(Bromomethyl)benzeneboronic acid pinacol ester
CAS 138500-85-3
Molecular Formula C13H18BBrO2
Formula Weight 297
MDL Number MFCD02179493
UN Number UN3261
Beilstein 7921808
Synonym 4-(Bromomethyl)phenylboronic acid pinacol ester; 2-[(4-Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAL1995403

  SDS
alfa aesar™
L1995403
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1995406

  SDS
alfa aesar™
L1995406
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 297
CAS 138500-85-3
Molecular Formula C13H18BBrO2
Assay 95%
Chemical Name or Material 4-(Bromomethyl)benzeneboronic acid pinacol ester
Health Hazard 1 Danger
Synonym 4-(Bromomethyl)phenylboronic acid pinacol ester; 2-[(4-Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD02179493
Quantity 1g
Melting Point 83° to 85°C
UN Number UN3261
Beilstein 7921808
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.