missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Bromomethyl)-7-methoxycoumarin, 97%, Thermo Scientific Chemicals

$168.52 - $168.52

Chemical Identifiers

CAS 35231-44-8
Molecular Formula C11H9BrO3
Molecular Weight (g/mol) 269.09
MDL Number MFCD00006869
InChI Key CTENSLORRMFPDH-UHFFFAOYSA-N
Synonym 4-bromomethyl-7-methoxycoumarin, 4-bromomethyl-7-methoxy-2h-chromen-2-one, br-mmc, ccris 7996, 4-bromomethyl-7-methoxychromen-2-one, 2h-1-benzopyran-2-one, 4-bromomethyl-7-methoxy, 4-bromomethyl-7-methoxy-2-oxo-2h-benzopyran, acmc-1agnd, 4-brommethyl-7-methoxy-2h-chromen-2-on, 4-bromomethyl-7-methoxy coumarin
PubChem CID 121894
IUPAC Name 4-(bromomethyl)-7-methoxychromen-2-one
SMILES COC1=CC=C2C(CBr)=CC(=O)OC2=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC365190010
SDS View Documents Promotion Details
Thermo Scientific Chemicals
365190010
1 g Glass bottle
Each for $168.52
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35231-44-8
269.09
CTENSLORRMFPDH-UHFFFAOYSA-N
121894
COC1=CC=C2C(CBr)=CC(=O)OC2=C1
C11H9BrO3
MFCD00006869
4-bromomethyl-7-methoxycoumarin, 4-bromomethyl-7-methoxy-2h-chromen-2-one, br-mmc, ccris 7996, 4-bromomethyl-7-methoxychromen-2-one, 2h-1-benzopyran-2-one, 4-bromomethyl-7-methoxy, 4-bromomethyl-7-methoxy-2-oxo-2h-benzopyran, acmc-1agnd, 4-brommethyl-7-methoxy-2h-chromen-2-on, 4-bromomethyl-7-methoxy coumarin
4-(bromomethyl)-7-methoxychromen-2-one

Specifications

35231-44-8
100.0
Beige or White to Yellow
96% min. (HPLC)
C11H9BrO3
1 g
Solubility in water: insoluble in water. Other solubilities: solubel in most common organic solvents
COC1=CC=C2C(CBr)=CC(=O)OC2=C1
269.09
269.08
Chunks and/or Powder
96.0
212.0°C to 217.0°C
Authentic
Glass bottle
MFCD00006869
4-bromomethyl-7-methoxycoumarin, 4-bromomethyl-7-methoxy-2h-chromen-2-one, br-mmc, ccris 7996, 4-bromomethyl-7-methoxychromen-2-one, 2h-1-benzopyran-2-one, 4-bromomethyl-7-methoxy, 4-bromomethyl-7-methoxy-2-oxo-2h-benzopyran, acmc-1agnd, 4-brommethyl-7-methoxy-2h-chromen-2-on, 4-bromomethyl-7-methoxy coumarin
CTENSLORRMFPDH-UHFFFAOYSA-N
4-(bromomethyl)-7-methoxychromen-2-one
121894
97%
4-(Bromomethyl)-7-methoxycoumarin
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 252-448-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only