Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester
Formula Weight 314.99
Synonym 4-Bromomethyl-3-fluorophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6284003

  SDS
alfa aesar™
H6284003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62840MD

  SDS
alfa aesar™
H62840MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 314.99
Chemical Name or Material 4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester
Physical Form Solid
Synonym 4-Bromomethyl-3-fluorophenylboronic acid pinacol ester
Quantity 1g
Assay 96%