missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromomethyl-2,6-dichloropyridine, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC433440050

 View more versions of this product

Catalog No. AC433440050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Bromomethyl-2, 6-dichloropyridine
Glass bottle
Yellow
5g
96.0
97%
MFCD00052637
XDKGMGJZRFOZRV-UHFFFAOYSA-N
4-(bromomethyl)-2,6-dichloropyridine
2781829
Crystalline Powder
Authentic
118.0°C (0.2 mmHg)
42.5°C to 43.2°C
175204-45-2
100.0
C6H4BrCl2N
4-bromomethyl-2,6-dichloropyridine, pyridine,4-bromomethyl-2,6-dichloro, 4-bromomethyl-2 6-dichloropyridine, 4-bromomethyl-2,6-dichloropyridin, 4-bromomethyl-2,6-dichloro-pyridine, 2,6-dichloro-4-bromomethyl pyridine, 4-bromomethyl-2,6-bis chloranyl pyridine
C1=C(C=C(N=C1Cl)Cl)CBr
240.909
241
97%

Chemical Identifiers

175204-45-2
240.909
XDKGMGJZRFOZRV-UHFFFAOYSA-N
2781829
C1=C(C=C(N=C1Cl)Cl)CBr
C6H4BrCl2N
MFCD00052637
4-bromomethyl-2,6-dichloropyridine, pyridine,4-bromomethyl-2,6-dichloro, 4-bromomethyl-2 6-dichloropyridine, 4-bromomethyl-2,6-dichloropyridin, 4-bromomethyl-2,6-dichloro-pyridine, 2,6-dichloro-4-bromomethyl pyridine, 4-bromomethyl-2,6-bis chloranyl pyridine
4-(bromomethyl)-2,6-dichloropyridine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents