Please login to your online account to display your discounted pricing

4-(Bromomethyl)-2,1,3-benzoxadiazole, Maybridge

$477.46 - $477.46

Chemical Identifiers

Chemical Name or Material 4-(Bromomethyl)-2, 1, 3-benzoxadiazole
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H5BrN2O
Formula Weight 213.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC09608CB

  SDS
fisher scientific company
CC09608CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC09608DA

  SDS
fisher scientific company
CC09608DA
Amber Glass Bottle 1g Each for $477.46
Description & Specifications

Specifications

Formula Weight 213.03
Quantity 250mg
Assay 95%
Chemical Name or Material 4-(Bromomethyl)-2, 1, 3-benzoxadiazole
Molecular Formula C7H5BrN2O
Assay Percent Range 95%
CAS 32863-30-2
Packaging Amber Glass Bottle