Please login to your online account to display your discounted pricing

4-(Bromomethyl)-2,1,3-benzoxadiazole, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC09608DA

 View more versions of this product

Catalog No. CC09608DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 213.03
Quantity 1g
Assay 95%
Assay Percent Range 95%
Chemical Name or Material 4-(Bromomethyl)-2, 1, 3-benzoxadiazole
Molecular Formula C7H5BrN2O
CAS 32863-30-2
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-(Bromomethyl)-2, 1, 3-benzoxadiazole
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H5BrN2O
Formula Weight 213.03