missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC64808DA

 View more versions of this product

Catalog No. CC64808DA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

335.12
97%
912569-72-3
4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole
96.5
Amber Glass Bottle
1g
100.0
C12H10BrF3N2O
MFCD09879947
97%

Chemical Identifiers

912569-72-3
100.0
C12H10BrF3N2O
MFCD09879947
97%
96.5
97%
335.12
4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger