missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole 97%, Maybridge

Chemical Identifiers

CAS 912569-72-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H10BrF3N2O
Formula Weight 335.12
MDL Number MFCD09879947
Chemical Name or Material 4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC64808CB SDS Fisher Scientific Company
CC64808CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC64808DA SDS Fisher Scientific Company
CC64808DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

912569-72-3
100.0
C12H10BrF3N2O
MFCD09879947
97%
96.5
97%
335.12
4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole

Specifications

912569-72-3
100.0
250mg
C12H10BrF3N2O
MFCD09879947
97%
96.5
Amber Glass Bottle
97%
335.12
4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger