Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromomandelic acid, 98+%

$64.65 - $783.50

Chemical Identifiers

Chemical Name or Material 4-Bromomandelic acid
CAS 6940-50-7
Molecular Formula C8H7BrO3
Formula Weight 231.05
MDL Number MFCD00004232
Beilstein 1567909
Merck Index 14,1424
Synonym 4-Bromo-alpha-hydroxyphenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1297806

  SDS
alfa aesar™
A1297806
5g Each for $64.65

AAA1297822

  SDS
alfa aesar™
A1297822
100g Each for $783.50

AAA1297814

  SDS
alfa aesar™
A1297814
25g Each for $250.91
Description & Specifications

Specifications

Formula Weight 231.05
Chemical Name or Material 4-Bromomandelic acid
TSCA TSCA
Melting Point 115° to 119°C
Quantity 5g
Assay ≥98%
Molecular Formula C8H7BrO3
MDL Number MFCD00004232
CAS 6940-50-7
Beilstein 1567909
Merck Index 14,1424
Synonym 4-Bromo-alpha-hydroxyphenylacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
4-Bromomandelic acid is used as a pharmaceutical intermediate. 4-bromomandelic acid is mainly used as an analytical reagent for zirconium.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.