missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromoisoquinoline 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 107090100

 View more versions of this product

Catalog No. AC107090100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Bromoisoquinoline
1532-97-4
100.0
Glass bottle
39° to 43°C
≥97.5%
10g
208.06
20, 384
98%
97.5
Authentic
White to light beige
Crystalline Solid
98%
C9H6BrN
MFCD00006904

Chemical Identifiers

1532-97-4
100.0
C9H6BrN
MFCD00006904
≥97.5%
97.5
98%
208.06
4-Bromoisoquinoline, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-244-8