Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromoisoquinoline, 98%

$61.64 - $709.69

Chemical Identifiers

Chemical Name or Material 4-Bromoisoquinoline
CAS 1532-97-4
Molecular Formula C9H6BrN
Formula Weight 208.06
MDL Number MFCD00006904
Beilstein 114431
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1595106

  SDS
alfa aesar™
A1595106
5g Each for $61.64

AAA1595114

  SDS
alfa aesar™
A1595114
25g Each for $216.97

AAA1595122

  SDS
alfa aesar™
A1595122
100g Each for $709.69
Description & Specifications

Specifications

Formula Weight 208.06
Melting Point 39° to 43°C
CAS 1532-97-4
Quantity 5g
MDL Number MFCD00006904
Boiling Point 285°C
Chemical Name or Material 4-Bromoisoquinoline
Beilstein 114431
Health Hazard 1 Warning
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C9H6BrN
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.