Please login to your online account to display your discounted pricing

4-Bromoisoindoline hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Bromoisoindoline hydrochloride
CAS 923590-95-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C8H8BrN·ClH
Formula Weight 234.52
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC435870010

  SDS
thermo fisher scientific
AC435870010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0% (HPLC)
Physical Form Crystalline Powder
Formula Weight 234.52
Quantity 1g
Color White to pale yellow
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 923590-95-8
Chemical Name or Material 4-Bromoisoindoline hydrochloride
Assay Percent Range 97%
Molecular Formula C8H8BrN·ClH
Packaging Glass bottle
Elemental Analysis 39.33 to 42.61 % C 5.73 to 6.21 % N
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.