missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromoimidazole, 98%

$68.64 - $261.97

Chemical Identifiers

CAS 2302-25-2
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.975
MDL Number MFCD00047021
InChI Key FHZALEJIENDROK-UHFFFAOYSA-N
Synonym 1h-imidazole, 4-bromo, 3kqm, 4 or 5-bromoimidazole, 4-bromo-1h-imidazole, 4-bromo-3h-imidazole, 4-bromo-imidazole, 4-bromoimidazole, 5-bromoimidazole, imidazole, 4 or 5-bromo-6ci,7ci, imidazole, 4-bromo
PubChem CID 96125
IUPAC Name 5-bromo-1H-imidazole
SMILES C1=C(NC=N1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6497206
View Documents
Alfa Aesar™
H6497206
5g Each for $68.64
 
AAH6497214
View Documents
Alfa Aesar™
H6497214
25g Each for $261.97
 
Description

Description

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2302-25-2
146.975
FHZALEJIENDROK-UHFFFAOYSA-N
96125
C1=C(NC=N1)Br
C3H3BrN2
MFCD00047021
1h-imidazole, 4-bromo, 3kqm, 4 or 5-bromoimidazole, 4-bromo-1h-imidazole, 4-bromo-3h-imidazole, 4-bromo-imidazole, 4-bromoimidazole, 5-bromoimidazole, imidazole, 4 or 5-bromo-6ci,7ci, imidazole, 4-bromo
5-bromo-1H-imidazole

Specifications

131°C to 135°C
2302-25-2
MFCD00047021
1615
Slightly soluble in water.
C1=C(NC=N1)Br
146.975
146.97
98%
5g
C3H3BrN2
UN2811
1h-imidazole, 4-bromo, 3kqm, 4 or 5-bromoimidazole, 4-bromo-1h-imidazole, 4-bromo-3h-imidazole, 4-bromo-imidazole, 4-bromoimidazole, 5-bromoimidazole, imidazole, 4 or 5-bromo-6ci,7ci, imidazole, 4-bromo
FHZALEJIENDROK-UHFFFAOYSA-N
5-bromo-1H-imidazole
96125
Solid
4-Bromoimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : NI3517450

TSCA : No

Recommended Storage : Ambient temperatures