missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Bromoheptanophenone, 97%

$39.69 - $147.46

Chemical Identifiers

CAS 99474-02-9
Molecular Formula C13H17BrO
Molecular Weight (g/mol) 269.182
MDL Number MFCD00276567
InChI Key LDVBFPQTLJKTLV-UHFFFAOYSA-N
Synonym 1-4-bromophenyl heptan-1-one, 1-bromo-4-n-heptanoylbenzene, maybridge1_002469
PubChem CID 2775719
IUPAC Name 1-(4-bromophenyl)heptan-1-one
SMILES CCCCCCC(=O)C1=CC=C(C=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1925203
View Documents
Alfa Aesar™
A1925203
1g
Each for $39.69
 
AAA1925206
View Documents
Alfa Aesar™
A1925206
5g
Each for $147.46
 
Description

Description

Specifications

Chemical Identifiers

99474-02-9
269.182
LDVBFPQTLJKTLV-UHFFFAOYSA-N
2775719
CCCCCCC(=O)C1=CC=C(C=C1)Br
C13H17BrO
MFCD00276567
1-4-bromophenyl heptan-1-one, 1-bromo-4-n-heptanoylbenzene, maybridge1_002469
1-(4-bromophenyl)heptan-1-one

Specifications

69°C to 72°C
99474-02-9
MFCD00276567
LDVBFPQTLJKTLV-UHFFFAOYSA-N
1-(4-bromophenyl)heptan-1-one
2775719
97%
1g
C13H17BrO
1-4-bromophenyl heptan-1-one, 1-bromo-4-n-heptanoylbenzene, maybridge1_002469
CCCCCCC(=O)C1=CC=C(C=C1)Br
269.182
269.19
4'-Bromoheptanophenone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications