missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromoheptane, tech. 85%, remainder mainly 3-bromoheptane

Chemical Identifiers

CAS 998-93-6
Molecular Formula C7H15Br
Molecular Weight (g/mol) 179.101
MDL Number MFCD00039182
InChI Key BNUTXEKPXPZAIT-UHFFFAOYSA-N
Synonym heptane, 4-bromo, 4-bromo-heptane, acmc-20ammr, 4-bromoheptane 5g
PubChem CID 70460
IUPAC Name 4-bromoheptane
SMILES CCCC(CCC)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6080706 View Documents Alfa Aesar™
H6080706
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

998-93-6
179.101
BNUTXEKPXPZAIT-UHFFFAOYSA-N
70460
CCCC(CCC)Br
C7H15Br
MFCD00039182
heptane, 4-bromo, 4-bromo-heptane, acmc-20ammr, 4-bromoheptane 5g
4-bromoheptane

Specifications

40°C (104°F)
5g
C7H15Br
UN1993
heptane, 4-bromo, 4-bromo-heptane, acmc-20ammr, 4-bromoheptane 5g
CCCC(CCC)Br
179.101
179.11
Technical
remainder mainly 3-bromoheptane
60°C to 62°C (18mmHg)
998-93-6
MFCD00039182
1697144
BNUTXEKPXPZAIT-UHFFFAOYSA-N
4-bromoheptane
70460
85%
4-Bromoheptane
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA