Please login to your online account to display your discounted pricing

4-Bromofluorobenzene 99%, ACROS Organics™


Quantity 10mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  107010100

 View more versions of this product

Catalog No. AC107010100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 175
Physical Form Liquid
Quantity 10mL
Color Clear colorless
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
TSCA TSCA
RTECS Number CY9033750
EINECS Number 207-300-2
Beilstein 05, 209
CAS Max % 100.0
Refractive Index 1.5260 to 1.5280 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 1 Warning
Assay Percent Range 99%
Linear Formula BrC6H4F
MDL Number MFCD00000342
Chemical Name or Material 4-Bromofluorobenzene
CAS 460-00-4
Name Note 99%
Molecular Formula C6H4BrF
CAS Min % 98.5
Infrared Spectrum Authentic
Synonym 1-Bromo-4-fluorobenzene
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Flammable liquid and vapor.

Specifications

Chemical Name or Material 4-Bromofluorobenzene
Name Note 99%
CAS 460-00-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H4BrF
Linear Formula BrC6H4F
Formula Weight 175
MDL Number MFCD00000342
Synonym 1-Bromo-4-fluorobenzene
Accessories

Warning

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents