missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromodiphenylamine, TCI America™

$29.03 - $99.31

Chemical Identifiers

CAS 54446-36-5
Molecular Formula C12H10BrN
Molecular Weight (g/mol) 248.123
MDL Number MFCD07779504
InChI Key CCIVUDMVXNBUCY-UHFFFAOYSA-N
Synonym N-(4-Bromophenyl)aniline, 4-Bromo-N-phenylaniline
PubChem CID 6425786
IUPAC Name 4-bromo-N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B39495G View Documents TCI America
B39495G
5g Each for $29.03
 
B394925G View Documents TCI America
B394925G
25g Each for $99.31
 
Specifications

Chemical Identifiers

54446-36-5
248.123
CCIVUDMVXNBUCY-UHFFFAOYSA-N
6425786
C1=CC=C(C=C1)NC2=CC=C(C=C2)Br
C12H10BrN
MFCD07779504
N-(4-Bromophenyl)aniline, 4-Bromo-N-phenylaniline
4-bromo-N-phenylaniline

Specifications

318°C
87°C
54446-36-5
MFCD07779504
CCIVUDMVXNBUCY-UHFFFAOYSA-N
4-bromo-N-phenylaniline
6425786
Crystal-Powder at 20°C
4-Bromodiphenylamine
White-Yellow
5g
C12H10BrN
N-(4-Bromophenyl)aniline, 4-Bromo-N-phenylaniline
C1=CC=C(C=C1)NC2=CC=C(C=C2)Br
248.123
248.12
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0061

TSCA : No