Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromocrotonic acid benzoylmethyl ester, 97%


Quantity 5g

Manufacturer: alfa aesar™  L1376506

 View more versions of this product

Catalog No. AAL1376506

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 283.13
Chemical Name or Material 4-Bromocrotonic acid benzoylmethyl ester
Molecular Formula C12H11BrO3
CAS 154561-38-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
MDL Number MFCD00236054
UN Number UN3335
Melting Point 89° to 91°C
Synonym 4-Bromocrotonic acid phenacyl ester; Phenacyl 4-bromocrotonate
Quantity 5g
Health Hazard 1 Warning
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.

Specifications

Chemical Name or Material 4-Bromocrotonic acid benzoylmethyl ester
CAS 154561-38-3
Molecular Formula C12H11BrO3
Formula Weight 283.13
MDL Number MFCD00236054
UN Number UN3335
Synonym 4-Bromocrotonic acid phenacyl ester; Phenacyl 4-bromocrotonate
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.