missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromochlorobenzene, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 106831000

Catalog No. AC106831000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Bromochlorobenzene
106-39-8
100.0
196.0°C
Authentic
Plastic bottle
BrC6H4Cl
100g
1.651
Solubility in water: 0.045g/L (25°C). Other solubilities: soluble in most organic solvents
ClC1=CC=C(Br)C=C1
191.45
191.45
99%
98.5
1.6510g/mL
>100°C
98.5% min. (GC)
C6H4BrCl
MFCD00000600
05, 209
4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
NHDODQWIKUYWMW-UHFFFAOYSA-N
1-bromo-4-chlorobenzene
7806
99%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-392-3

RTECSNumber : CY9006000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only