Please login to your online account to display your discounted pricing

4-Bromochlorobenzene 99%, ACROS Organics™


Quantity 50g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC106830500

 View more versions of this product

Catalog No. AC106830500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 191.45
Physical Form Crystalline Powder
Quantity 50g
Color White to almost white
Beilstein 05, 209
RTECS Number CY9006000
TSCA TSCA
Melting Point 64° to 67°C
Density 1.651g/mL
EINECS Number 203-392-3
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99%
Molecular Formula C6H4BrCl
MDL Number MFCD00000600
Chemical Name or Material 4-Bromochlorobenzene
Assay Percent Range 99%
CAS Min % 98.5
CAS 106-39-8
Linear Formula BrC6H4Cl
Packaging Plastic bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Bromochlorobenzene
Name Note 99%
CAS 106-39-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H4BrCl
Linear Formula BrC6H4Cl
Formula Weight 191.45
MDL Number MFCD00000600
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents