missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromobutyryl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1209406

 View more versions of this product

Catalog No. AAA1209406



Description

Description

4-Bromobutyryl chloride is used as a precursor for the synthesis of carolic acid by reacting with ethoxymagnesiomalonic ester. It is widely utilized as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobutyryl chloride is used as a precursor for the synthesis of carolic acid by reacting with ethoxymagnesiomalonic ester. It is widely utilized as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Remove all sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

4-Bromobutyryl chloride
1.6
91°C (195°F)
1.492
MFCD00000753
UN3265
Moisture sensitive
Miscible with organic solvents. Immiscible with water.
C(CC(=O)Cl)CBr
185.445
185.45
927-58-2
102°C to 103°C (40 mmHg)
C4H6BrClO
Br(CH2)3COCl
5 g
1743026
4-bromobutyryl chloride, butanoyl chloride, 4-bromo, g-bromobutyroyl chloride, butyryl chloride, 4-bromo, 4-bromobutyrylchloride, bromobutryl chloride, bromobutyryl chloride, acmc-20ammt, 4-bromobutanoylchloride, 4-bromobutyroylchloride
LRTRXDSAJLSRTG-UHFFFAOYSA-N
4-bromobutanoyl chloride
70225
97%

Chemical Identifiers

927-58-2
185.445
LRTRXDSAJLSRTG-UHFFFAOYSA-N
70225
C(CC(=O)Cl)CBr
C4H6BrClO
MFCD00000753
4-bromobutyryl chloride, butanoyl chloride, 4-bromo, g-bromobutyroyl chloride, butyryl chloride, 4-bromo, 4-bromobutyrylchloride, bromobutryl chloride, bromobutyryl chloride, acmc-20ammt, 4-bromobutanoylchloride, 4-bromobutyroylchloride
4-bromobutanoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 213-154-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only