missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromobutyronitrile 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 106800050

 View more versions of this product

Catalog No. AC106800050


 

Specifications Description & Specifications

Specifications

≥96.0%
Liquid
Clear colorless to yellow
96.0
4-Bromobutanenitrile
Br(CH2)3CN
C4H6BrN
5332-06-9
Authentic
Glass bottle
148
5mL
02, 283
100.0
97%
97%
4-Bromobutyronitrile
MFCD00001971
1.4770 to 1.4790 (20°C, 589nm)

Chemical Identifiers

5332-06-9
100.0
C4H6BrN
148
4-Bromobutanenitrile
≥96.0%
96.0
97%
Br(CH2)3CN
MFCD00001971
4-Bromobutyronitrile, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 226-236-6

RTECSNumber : ET8925000

TSCA : TSCA