Learn More
4-Bromobutyronitrile, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1067409
Description
4-bromobutyronitrile is used as a precursor for the preparation of cylopropyl cyanide. Its derivative, 4-Bromo-2,2-Diphenyl Butyronitrile is used as an intermediate for loperamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-bromobutyronitrile is used as a precursor for the preparation of cylopropyl cyanide. Its derivative, 4-Bromo-2,2-Diphenyl Butyronitrile is used as an intermediate for loperamide.
Solubility
Miscible with toluene and benzene. Immiscible with water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Specifications
4-Bromobutyronitrile | |
1.491 | |
103°C (217°F) | |
C4H6BrN | |
MFCD00001971 | |
UN3276 | |
4-bromobutyronitrile, butanenitrile, 4-bromo, 1-bromo-3-cyanopropane, 3-bromopropyl cyanide, 3-cyanopropyl bromide, gamma-bromobutyronitrile, butyronitrile, 4-bromo, usaf do-6, 4-bromo butyronitrile, trimethylene bromocyanide | |
CQPGDDAKTTWVDD-UHFFFAOYSA-N | |
4-bromobutanenitrile | |
21412 | |
97% |
5332-06-9 | |
204°C to 206°C | |
Odorless | |
1.478 | |
10 g | |
956577 | |
Miscible with toluene and benzene. Immiscible with water. | |
C(CC#N)CBr | |
148.00 | |
148.01 |
Chemical Identifiers
5332-06-9 | |
148.00 | |
CQPGDDAKTTWVDD-UHFFFAOYSA-N | |
21412 | |
C(CC#N)CBr |
C4H6BrN | |
MFCD00001971 | |
4-bromobutyronitrile, butanenitrile, 4-bromo, 1-bromo-3-cyanopropane, 3-bromopropyl cyanide, 3-cyanopropyl bromide, gamma-bromobutyronitrile, butyronitrile, 4-bromo, usaf do-6, 4-bromo butyronitrile, trimethylene bromocyanide | |
4-bromobutanenitrile |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 226-236-6
RTECSNumber : ET8925000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only