Please login to your online account to display your discounted pricing

4-Bromobutyl acetate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Bromobutyl acetate
CAS 4753-59-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H11BrO2
Linear Formula CH3CO2(CH2)4Br
Formula Weight 195.06
MDL Number MFCD00000263
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC210990500

  SDS
thermo fisher scientific
AC210990500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC210990100

  SDS
thermo fisher scientific
210990100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 195.06
Quantity 50g
Color Clear colorless to slightly yellow
Density 1.3480g/mL
Beilstein 02, IV, 147
CAS Max % 100.0
Chemical Name or Material 4-Bromobutyl acetate
Assay Percent Range 98%
Molecular Formula C6H11BrO2
Infrared Spectrum Authentic
MDL Number MFCD00000263
CAS 4753-59-7
CAS Min % 97.5
Packaging Glass bottle
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Linear Formula CH3CO2(CH2)4Br
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.