missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromobutyl acetate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 210990100

 View more versions of this product

Catalog No. AC210990100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
195.06
Clear colorless to slightly yellow
100.0
1.4590 to 1.4610 (20°C, 589nm)
C6H11BrO2
97.5
4753-59-7
98%
Liquid
10g
02, IV, 147
Glass bottle
4-Bromobutyl acetate
MFCD00000263
Authentic
CH3CO2(CH2)4Br

Chemical Identifiers

4753-59-7
100.0
C6H11BrO2
195.06
4-Bromobutyl acetate
97.5
98%
CH3CO2(CH2)4Br
MFCD00000263
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.