missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromobutyl acetate, 98+%

Manufacturer:  Alfa Aesar™ A1563230

 View more versions of this product

Catalog No. AAA1563230



Specifications

Specifications

4-Bromobutyl acetate
96°C to 97°C (15mmHg)
4753-59-7
MFCD00000263
4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide
CC(=O)OCCCCBr
195.056
195.07
109°C (228°F)
250g
C6H11BrO2
1749695
UOABIRUEGSGTSA-UHFFFAOYSA-N
4-bromobutyl acetate
78491
≥98%

Chemical Identifiers

4753-59-7
195.056
UOABIRUEGSGTSA-UHFFFAOYSA-N
78491
CC(=O)OCCCCBr
C6H11BrO2
MFCD00000263
4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide
4-bromobutyl acetate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents