missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromobutyl acetate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1563209
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Bromobutyl acetate | |
1.348 | |
109°C (228°F) | |
1.46 | |
10 g | |
4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide | |
CC(=O)OCCCCBr | |
195.056 | |
195.07 |
4753-59-7 | |
96°C to 97°C (15 mmHg) | |
C6H11BrO2 | |
MFCD00000263 | |
1749695 | |
UOABIRUEGSGTSA-UHFFFAOYSA-N | |
4-bromobutyl acetate | |
78491 | |
≥98% |
Chemical Identifiers
4753-59-7 | |
195.056 | |
UOABIRUEGSGTSA-UHFFFAOYSA-N | |
78491 | |
CC(=O)OCCCCBr |
C6H11BrO2 | |
MFCD00000263 | |
4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide | |
4-bromobutyl acetate |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 225-277-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only