missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Bromobutoxy)benzene, 97%

Manufacturer:  Alfa Aesar™ A1495606

 View more versions of this product

Catalog No. AAA1495606



Description

Description

Applications
(4-Bromobutoxy)benzene is used to produce 5-phenoxy-valeric acid with carbon dioxide and reagent Mg.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

(4-Bromobutoxy)benzene
Odorless
40°C to 43°C
1200-03-9
MFCD00000262
4-bromobutoxy benzene, 4-phenoxybutyl bromide, 4-bromobutyl phenyl ether, benzene, 4-bromobutoxy, 1-bromo-4-phenoxybutane, phenoxybutyl bromide, 4-phenoxybutylbromide, benzene, bromobutoxy, 4-bromo-1-phenoxybutane, 4-brombutoxy benzol
C1=CC=C(C=C1)OCCCCBr
229.117
229.12
>110°C (230°F)
153°C to 154°C (18mmHg)
5g
C10H13BrO
2044374
QBLISOIWPZSVIK-UHFFFAOYSA-N
4-bromobutoxybenzene
70986
97%

Chemical Identifiers

1200-03-9
229.117
QBLISOIWPZSVIK-UHFFFAOYSA-N
70986
C1=CC=C(C=C1)OCCCCBr
C10H13BrO
MFCD00000262
4-bromobutoxy benzene, 4-phenoxybutyl bromide, 4-bromobutyl phenyl ether, benzene, 4-bromobutoxy, 1-bromo-4-phenoxybutane, phenoxybutyl bromide, 4-phenoxybutylbromide, benzene, bromobutoxy, 4-bromo-1-phenoxybutane, 4-brombutoxy benzol
4-bromobutoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents