missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Bromobiphenyl-3-carboxylic acid, 95%

$90.19 - $289.38

Chemical Identifiers

CAS 885951-66-6
Molecular Formula C13H9BrO2
Formula Weight 277.12
MDL Number MFCD01318398
Synonym 3-(4-Bromophenyl)benzoic acid
Chemical Name or Material 4'-Bromobiphenyl-3-carboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5176203 SDS Alfa Aesar™
H5176203
1g Each for $90.19
AAH5176206 SDS Alfa Aesar™
H5176206
5g Each for $289.38
Specifications Description & Specifications

Chemical Identifiers

885951-66-6
277.12
3-(4-Bromophenyl)benzoic acid
C13H9BrO2
MFCD01318398
4'-Bromobiphenyl-3-carboxylic acid

Specifications

885951-66-6
277.12
3-(4-Bromophenyl)benzoic acid
4'-Bromobiphenyl-3-carboxylic acid
C13H9BrO2
MFCD01318398
1g
95%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning