Please login to your online account to display your discounted pricing

4-Bromobenzylamine hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Bromobenzylamine hydrochloride
Name Note 98%
CAS 26177-44-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7 H8 Br N . H Cl
Linear Formula BrC6H4CH2NH2·HCl
Formula Weight 222.52
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC194650050

  SDS
thermo fisher scientific
194650050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 222.52
Quantity 5g
Color Off-white to beige
Beilstein 12, 1075
EINECS Number 247-503-3
Melting Point 272° to 276°C
CAS Max % 100.0
Molecular Formula C7 H8 Br N . H Cl
Name Note 98%
Assay Percent Range 98%
CAS 26177-44-6
CAS Min % 97.5
Chemical Name or Material 4-Bromobenzylamine hydrochloride
Linear Formula BrC6H4CH2NH2·HCl
MDL Number MFCD00012860
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.