missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromobenzyl Bromide, TCI America™

$22.92 - $262.78

Chemical Identifiers

CAS 589-15-1
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000179
InChI Key YLRBJYMANQKEAW-UHFFFAOYSA-N
Synonym 4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide
PubChem CID 68527
IUPAC Name 1-bromo-4-(bromomethyl)benzene
SMILES C1=CC(=CC=C1CBr)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B100225G View Documents TCI America
B100225G
25g Each for $22.92
 
B1002500G View Documents TCI America
B1002500G
500g Each for $262.78
 
B1002100G View Documents TCI America
B1002100G
100g Each for $64.17
 
Specifications

Chemical Identifiers

589-15-1
249.933
YLRBJYMANQKEAW-UHFFFAOYSA-N
68527
C1=CC(=CC=C1CBr)Br
C7H6Br2
MFCD00000179
4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide
1-bromo-4-(bromomethyl)benzene

Specifications

124°C
61°C
589-15-1
MFCD00000179
4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide
C1=CC(=CC=C1CBr)Br
249.933
249.93
≥98.0% (GC)
White
25g
C7H6Br2
3261
YLRBJYMANQKEAW-UHFFFAOYSA-N
1-bromo-4-(bromomethyl)benzene
68527
Crystal-Powder at 20°C
4-Bromobenzyl Bromide
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4330

TSCA : Yes