missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Bromobenzyl Bromide 98.0+%, TCI America™
Supplier: TCI America B1002100G
Specifications
4-Bromobenzyl Bromide | |
61°C | |
124°C | |
MFCD00000179 | |
3261 | |
YLRBJYMANQKEAW-UHFFFAOYSA-N | |
1-bromo-4-(bromomethyl)benzene | |
68527 | |
≥98.0% (GC) |
589-15-1 | |
White | |
C7H6Br2 | |
100 g | |
4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide | |
C1=CC(=CC=C1CBr)Br | |
249.933 | |
249.93 | |
Crystalline Powder |
Chemical Identifiers
589-15-1 | |
249.933 | |
YLRBJYMANQKEAW-UHFFFAOYSA-N | |
68527 | |
C1=CC(=CC=C1CBr)Br |
C7H6Br2 | |
MFCD00000179 | |
4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide | |
1-bromo-4-(bromomethyl)benzene |
Safety and Handling
EINECSNumber : (3)-4330
TSCA : Yes