missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromobenzyl alcohol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 181770500

 View more versions of this product

Catalog No. AC181770500


 

Specifications Description & Specifications

Specifications

4-Bromobenzyl alcohol
873-75-6
98.5
Glass bottle
75° to 78°C
Crystals or Crystalline Powder
99%
BrC6H4CH2OH
MFCD00004650
99%
Authentic
100.0
White to slightly beige
50g
≥98.5%
C7H7BrO
187.04
06, 446

Chemical Identifiers

873-75-6
100.0
C7H7BrO
187.04
4-Bromobenzyl alcohol, 99%
98.5
99%
BrC6H4CH2OH
MFCD00004650
≥98.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 212-851-7