missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromobenzotrifluoride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC166671000

 View more versions of this product

Catalog No. AC166671000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Bromobenzotrifluoride
48°C
Glass Bottle
1.6
Colorless to Yellow
402-43-7
100.0
C7H4BrF3
MFCD00000398
XLQSXGGDTHANLN-UHFFFAOYSA-N
1-bromo-4-(trifluoromethyl)benzene
67872
Liquid
1.6000g/mL
Authentic
1.4720 to 1.4740
154.0°C to 155.0°C
100mL
98.5
98.5% min. (GC)
BrC6H4CF3
1-bromo-4-trifluoromethyl benzene
C1=CC(=CC=C1C(F)(F)F)Br
225.01
225.01
99%

Chemical Identifiers

402-43-7
225.01
XLQSXGGDTHANLN-UHFFFAOYSA-N
67872
C1=CC(=CC=C1C(F)(F)F)Br
C7H4BrF3
MFCD00000398
1-bromo-4-trifluoromethyl benzene
1-bromo-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-943-6

SDS Documents