Please login to your online account to display your discounted pricing

4-Bromobenzophenone 97%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  106721000

 View more versions of this product

Catalog No. AC106721000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 261.11
Quantity 100g
Color White to light beige
Beilstein 07, IV, 1378
Melting Point 78° to 82°C
RTECS Number DJ0350000
EINECS Number 202-024-9
CAS Max % 100.0
CAS Min % 96.0
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00000103
Molecular Formula C13H9BrO
Assay Percent Range 97%
Linear Formula BrC6H4COC6H5
CAS 90-90-4
Chemical Name or Material 4-Bromobenzophenone
Name Note 97%
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Bromobenzophenone
Name Note 97%
CAS 90-90-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H9BrO
Linear Formula BrC6H4COC6H5
Formula Weight 261.11
MDL Number MFCD00000103
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents