Please login to your online account to display your discounted pricing

4-Bromobenzophenone 97%, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  106720250

 View more versions of this product

Catalog No. AC106720250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 261.11
Physical Form Crystalline Powder
Quantity 25g
Color White to light beige
RTECS Number DJ0350000
EINECS Number 202-024-9
Melting Point 78° to 82°C
Beilstein 07, IV, 1378
CAS Min % 96.0
CAS Max % 100.0
Packaging Plastic bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula BrC6H4COC6H5
Molecular Formula C13H9BrO
Assay Percent Range 97%
Name Note 97%
Chemical Name or Material 4-Bromobenzophenone
CAS 90-90-4
MDL Number MFCD00000103
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 4-Bromobenzophenone
Name Note 97%
CAS 90-90-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H9BrO
Linear Formula BrC6H4COC6H5
Formula Weight 261.11
MDL Number MFCD00000103
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents