Please login to your online account to display your discounted pricing

4-Bromobenzonitrile, 98%, ACROS Organics™


Quantity 10g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  106710100

 View more versions of this product

Catalog No. AC106710100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder or Flakes
Formula Weight 182.02
Color White to light yellow or flakes
Quantity 10g
Packaging Glass Bottle
RTECS Number DI2460000
EINECS Number 210-764-9
Melting Point 111° to 114°C
CAS Max % 100.0
CAS 623-00-7
Molecular Formula C7H4BrN
Name Note 98%
MDL Number MFCD00001811
Assay Percent Range 98%
CAS Min % 97.5
Linear Formula BrC6H4CN
Chemical Name or Material 4-Bromobenzonitrile
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.

Specifications

Chemical Name or Material 4-Bromobenzonitrile
Name Note 98%
CAS 623-00-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4BrN
Linear Formula BrC6H4CN
Formula Weight 182.02
MDL Number MFCD00001811
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents