missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromobenzonitrile, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC106710500

 View more versions of this product

Catalog No. AC106710500


 

Specifications Description & Specifications

Specifications

Powder or Flakes
≥97.5%
50g
100.0
4-Bromobenzonitrile
623-00-7
C7H4BrN
98%
Authentic
White to light yellow or flakes
182.02
111° to 114°C
Plastic bottle
BrC6H4CN
98%
MFCD00001811
97.5

Chemical Identifiers

623-00-7
100.0
C7H4BrN
182.02
4-Bromobenzonitrile, 98%
97.5
98%
BrC6H4CN
MFCD00001811
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 210-764-9

RTECSNumber : DI2460000