Please login to your online account to display your discounted pricing

4-Bromobenzoic hydrazide 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC386460010

 View more versions of this product

Catalog No. AC386460010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 215.05
Physical Form Powder
Quantity 1g
Color Light beige
Melting Point 164° to 168°C
CAS Max % 100.0
Linear Formula BrC6H4C(O)NHNH2
Chemical Name or Material 4-Bromobenzoic hydrazide
Infrared Spectrum Authentic
MDL Number MFCD00007602
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Min % 97.5
Packaging Glass bottle
CAS 5933-32-4
Synonym 4-Bromobenzhydrazide
Molecular Formula C7H7BrN2O
Assay Percent Range 98%

Specifications

Chemical Name or Material 4-Bromobenzoic hydrazide
CAS 5933-32-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7BrN2O
Linear Formula BrC6H4C(O)NHNH2
Formula Weight 215.05
MDL Number MFCD00007602
Synonym 4-Bromobenzhydrazide
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents